accessibility menu, dialog, popup

UserName

CAS RN 41253-21-8

IUPAC Name: sodium 4H-1,2,4-triazol-4-ide
Synonyms: sodium 4H-1,2,4-triazol-4-ide
Molecular Weight (g/mol): 91.05
Molecular Formula: C2H2N3Na
InChi Key: NVMNEWNGLGACBB-UHFFFAOYSA-N
SMILES: [Na+].[N-]1C=NN=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)