missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1,2,4-Triazolide 96.0+%, TCI America™

Catalog No. S052225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S052225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S052225G Supplier TCI America Supplier No. S052225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41253-21-8
Molecular Formula C2H2N3Na
Molecular Weight (g/mol) 91.049
MDL Number MFCD00044437
InChI Key NVMNEWNGLGACBB-UHFFFAOYSA-N
Synonym sodium 1,2,4-triazol-1-ide, 1,2,4-triazolylsodium, 1,2,4-triazole sodium derivative, 1,2,4-triazole sodium salt, sodium 1,2,4-triazolide, 1h-1,2,4-triazole, sodium salt, sodium 1h-1,2,4-triazolate, 1,2,4-triazole sodium, sodium 1h-1,2,4-triazolide, 1,2,4-triazole, sodium salt
PubChem CID 4649906
IUPAC Name sodium;2,4-diaza-1-azanidacyclopenta-2,4-diene
SMILES C1=NC=N[N-]1.[Na+]

Specifications

Melting Point 327°C
Color White
Quantity 25 g
Formula Weight 91.05
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1,2,4-Triazolide