accessibility menu, dialog, popup

UserName

CAS RN 4064-06-6

IUPAC Name: {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
Synonyms: {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: POORJMIIHXHXAV-UHFFFAOYNA-N
SMILES: CC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)