missing translation for 'onlineSavingsMsg'
Learn More

Diacetone-D-galactose, 95%

Catalog No. AAB2489914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2489914 25 g
AAB2489906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2489914 Supplier Thermo Scientific Chemicals Supplier No. B2489914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4064-06-6 | C12H20O6 | 260.29 g/mol

Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

Solubility
Soluble in Acetone, Chloroform and Methanol.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.29
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
PubChem CID 70793572
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydro-3aH-di[1,3]dioxolo[4,5-a
SMILES CC1(OC2C(OC3C(C2O1)OC(O3)(C)C)CO)C

Specifications

Density 1.143
Quantity 25 g
Solubility Information Soluble in Acetone,Chloroform and Methanol.
Formula Weight 260.29
Percent Purity 97%
Chemical Name or Material Diacetone-D-galactose
EINECSNumber 223-771-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only