missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%

Catalog No. AC115240250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115240250 25 g Glass bottle
AC115240050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115240250 Supplier Thermo Scientific Chemicals Supplier No. 115240250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4064-06-6 | C12H20O6 | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 122.0°C
Color Yellow
Density 1.1400g/mL
Boiling Point 117.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4668 to 1.4688
Quantity 25 g
Specific Gravity 1.14
Specific Rotation Condition − 57.50 (20.00°C c=3,CHCl3)
Specific Rotation -57.5
Synonym n-4-methylphenyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolylacetamide, cbmicro_030789, n-p-tolyl-2,2-diphenylacetamide, 2,2-diphenyl-n-p-tolyl acetamide, nbeta-z-nin-boc-l-beta-homotryptophan, 2,2-diphenyl-n-p-tolyl-acetamide, n-4-methylphenyl-2,2-diphenyl-acetamide, n∼1∼-4-methylphenyl-2,2-diphenylacetamide
SMILES CC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
IUPAC Name {4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
PubChem CID 344833
Formula Weight 260.28
Percent Purity 97%
Physical Form Viscous Paste
Chemical Name or Material 1, 2:3, 4-Di-O-isopropylidene-D-galactopyranose, 97%
Show More Show Less
EINECSNumber 223-771-7
TSCA TSCA

RUO – Research Use Only