accessibility menu, dialog, popup

UserName

ClOOO

CAS RN 4053-08-1

IUPAC Name: 10-chloro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 10-chloro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 232.62
Molecular Formula: C12H5ClO3
InChi Key: UJEUBSWHCGDJQU-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)