missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,8-naphthalic anhydride, 94%

Catalog No. AAL0550836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0550836 500 g
AAL0550814 25 g
AAL0550822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0550836 Supplier Thermo Scientific Chemicals Supplier No. L0550836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4053-08-1 | C12H5ClO3 | 232.62 g/mol

4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.

Chemical Identifiers

CAS 4053-08-1
Molecular Formula C12H5ClO3
Molecular Weight (g/mol) 232.62
MDL Number MFCD00006928
InChI Key UJEUBSWHCGDJQU-UHFFFAOYSA-N
IUPAC Name 10-chloro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione

Specifications

Melting Point 208°C to 214°C
Quantity 500 g
Beilstein 178072
Sensitivity Moisture sensitive
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
Solubility Information Hydrolyzes in water.
SMILES ClC1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23
PubChem CID 70793572
Formula Weight 232.63
Percent Purity 94%
Chemical Name or Material 4-Chloro-1,8-naphthalic anhydride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-760-7
RTECSNumber QL6127295
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only