accessibility menu, dialog, popup

UserName

NHNNNNH2NH2H2O

CAS RN 402846-48-4

IUPAC Name: 7H-purine-2,6-diamine hydrate
Synonyms: 2,6-diaminopurine hydrate
Molecular Weight (g/mol): 168.16
Molecular Formula: C5H8N6O
InChi Key: UKADKJJNEZZNGH-UHFFFAOYSA-N
SMILES: O.NC1=NC(N)=C2NC=NC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results