missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopurine Monohydrate 97.0+%, TCI America™

Catalog No. D16255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16255G 5 g
D162525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16255G Supplier TCI America Supplier No. D16255G
Only null left

Chemical Identifiers

CAS 402846-48-4
Molecular Formula C5H8N6O
Molecular Weight (g/mol) 168.16
MDL Number MFCD00213668
InChI Key UKADKJJNEZZNGH-UHFFFAOYSA-N
PubChem CID 19092948
IUPAC Name 7H-purine-2,6-diamine;hydrate
SMILES C1=NC2=C(N1)C(=NC(=N2)N)N.O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 150.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminopurine Monohydrate
RTECSNumber UO7524000
TSCA Yes