accessibility menu, dialog, popup

UserName

CAS RN 40235-68-5

IUPAC Name: 3-chloro-2-oxopropyl acetate
Synonyms: 3-chloro-2-oxopropyl acetate
Molecular Weight (g/mol): 150.56
Molecular Formula: C5H7ClO3
InChi Key: LCMPIWIWBGHBOY-UHFFFAOYSA-N
SMILES: CC(=O)OCC(=O)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results