accessibility menu, dialog, popup

UserName

HOBOHO

CAS RN 40138-16-7

IUPAC Name: (2-formylphenyl)boronic acid
Synonyms: 2-formylphenylboronic acid
Molecular Weight (g/mol): 149.94
Molecular Formula: C7H7BO3
InChi Key: DGUWACLYDSWXRZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C=O
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)