missing translation for 'onlineSavingsMsg'
Learn More

2-Formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B18731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18731G Supplier TCI America Supplier No. B18731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Formylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 40138-16-7
Melting Point 125°C
Molecular Formula C7H7BO3
Quantity 1 g
MDL Number MFCD00151822
Synonym 2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
InChI Key DGUWACLYDSWXRZ-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC=C1C=O
IUPAC Name (2-formylphenyl)boronic acid
Molecular Weight (g/mol) 149.94
PubChem CID 292189
Formula Weight 149.94
Physical Form Crystalline Powder
Show More Show Less