accessibility menu, dialog, popup

UserName

H3CH2N(S)OHOH

CAS RN 40137-22-2

IUPAC Name: [(2S)-2,3-dihydroxypropyl](methyl)azanium
Synonyms: [(2S)-2,3-dihydroxypropyl](methyl)azanium
Molecular Weight (g/mol): 106.14
Molecular Formula: C4H12NO2
InChi Key: WOMTYMDHLQTCHY-BYPYZUCNSA-O
SMILES: C[NH2+]C[C@H](O)CO
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)