missing translation for 'onlineSavingsMsg'
Learn More

3-Methylamino-1,2-propanediol 98.0+%, TCI America™

Catalog No. M124325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M124325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M124325G Supplier TCI America Supplier No. M124325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40137-22-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00082732
InChI Key WOMTYMDHLQTCHY-UHFFFAOYSA-N
Synonym 3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
PubChem CID 2734122
IUPAC Name 3-(methylamino)propane-1,2-diol
SMILES CNCC(CO)O

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 g
UN Number 2735
Formula Weight 105.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylamino-1,2-propanediol
EINECSNumber (2)-3971
TSCA No