accessibility menu, dialog, popup

UserName

FFFBr

CAS RN 401-78-5

IUPAC Name: 1-bromo-3-(trifluoromethyl)benzene
Synonyms: 1-bromo-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: NNMBNYHMJRJUBC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(Br)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)