accessibility menu, dialog, popup

UserName

CAS RN 40045-50-9

SNSNNSNH2NOO

CAS RN 40045-50-9

IUPAC Name: 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine
Synonyms: 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 261.29
Molecular Formula: C5H3N5O2S3
InChi Key: NQQBNZBOOHHVQP-UHFFFAOYSA-N
SMILES: NC1=NN=C(SC2=NC=C(S2)[N+]([O-])=O)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results