missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole 96.0+%, TCI America™

Catalog No. A294020MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A294020MG 20 mg
A2940100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A294020MG Supplier TCI America Supplier No. A294020MG
Only null left

Chemical Identifiers

CAS 40045-50-9
Molecular Formula C5H3N5O2S3
Molecular Weight (g/mol) 261.292
MDL Number MFCD01927019
InChI Key NQQBNZBOOHHVQP-UHFFFAOYSA-N
Synonym SU 3327
PubChem CID 11837140
IUPAC Name 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine
SMILES C1=C(SC(=N1)SC2=NN=C(S2)N)[N+](=O)[O-]

Specifications

Melting Point 160°C
Quantity 20 mg
Formula Weight 261.29
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole
TSCA No
Recommended Storage Freezer