accessibility menu, dialog, popup

UserName

CAS RN 3992-42-5

IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
Synonyms: 2'-deoxycytidine hcl
Molecular Weight (g/mol): 263.68
Molecular Formula: C9H14ClN3O4
InChi Key: LTKCXZGFJFAPLY-VOBHXVAMNA-N
SMILES: Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)