missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxycytidine hydrochloride, Analytical Standard, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014137
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014137 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014137 Supplier Merck Emd Millipore Supplier No. D80061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Poly Bottle
CAS 3992-42-5
Physical Form Neat
Linear Formula C9H13N3O4 · HCl
MDL Number MFCD00012840
Synonym 1-(2-Deoxy-beta-D-ribofuranosyl)cytosine hydrochloride; Cytosine deoxyriboside hydrochloride
Quantity 1 g
Formula Weight 263.68
Percent Purity ≥99%
Grade Analytical Standard