accessibility menu, dialog, popup

UserName

CAS RN 399-76-8

IUPAC Name: 5-fluoro-1H-indole-2-carboxylate
Synonyms: 5-fluoro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 178.14
Molecular Formula: C9H5FNO2
InChi Key: WTXBRZCVLDTWLP-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(F)=CC=C2N1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)