accessibility menu, dialog, popup

UserName

H2N(S)OHOHN(R)(S)NH2(R)O(R)O(R)H2N(S)OH(S)OH(R)OH(S)OH(S)O(S)O(R)OH(S)OH(S)H2N(R)HOOHSOHOOOHSOHOO

CAS RN 39831-55-5

IUPAC Name: (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid)
Synonyms: amikacin; bis(sulfuric acid)
Molecular Weight (g/mol): 781.75
Molecular Formula: C22H47N5O21S2
InChi Key: FXKSEJFHKVNEFI-GCZBSULCSA-N
SMILES: OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Type 
  • (4)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results