missing translation for 'onlineSavingsMsg'
Learn More

Amikacin disulfate salt, Thermo Scientific Chemicals

Catalog No. AC455190050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455190050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455190050 Supplier Thermo Scientific Chemicals Supplier No. 455190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39831-55-5
Molecular Formula C22H47N5O21S2
Molecular Weight (g/mol) 781.75
MDL Number MFCD00167475,MFCD00167475
InChI Key FXKSEJFHKVNEFI-GCZBSULCSA-N
PubChem CID 134129615
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4S,5S)-5-amino-2-[(2S,3S,4S,5S,6S)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3S,4S,5S,6S)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
SMILES OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O

Specifications

Melting Point 220.0°C to 230.0°C
pH 2 to 4 (1% aq. soln.)
Loss on Drying 13% max. (110°C, 3 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: ca 50g/L.
Formula Weight 781.76
Chemical Name or Material Amikacin disulfate salt
EINECSNumber 254-648-6

RUO – Research Use Only