missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amikacin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 39831-55-5 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C22H43N5O13 · 2H2 SO4 |
| MDL Number | MFCD00167475 |
| Synonym | Amikacin disulfate salt |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 781.76 |
| Grade | Certified Reference Material |