accessibility menu, dialog, popup

UserName

CAS RN 39718-32-6

IUPAC Name: 1,4-difluoro-2-isocyanatobenzene
Synonyms: 1,4-difluoro-2-isocyanatobenzene
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: SNHIIFOXCRYGGY-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)