missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenyl Isocyanate 98.0+%, TCI America™

Catalog No. D36955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36955G 5 g
D36951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36955G Supplier TCI America Supplier No. D36955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39718-32-6
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00013843
InChI Key SNHIIFOXCRYGGY-UHFFFAOYSA-N
Synonym 2,5-difluorophenyl isocyanate, 2,5-difluorophenylisocyanate, benzene, 1,4-difluoro-2-isocyanato, isocyanic acid 2,5-difluorophenyl ester, 2,5-difluorobenzenisocyanate, pubchem5022, acmc-1ahno, 2,5-difluoro-phenyl-isocyanate, benzene,1,4-difluoro-2-isocyanato
PubChem CID 2733266
IUPAC Name 1,4-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C=C1F)N=C=O)F

Specifications

Color Yellow
Boiling Point 54°C
Quantity 5 g
UN Number 3080
Formula Weight 155.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator