accessibility menu, dialog, popup

UserName

BrSBrBrBr

CAS RN 3958-03-0

IUPAC Name: tetrabromothiophene
Synonyms: thiophene, tetrabromo-
Molecular Weight (g/mol): 399.72
Molecular Formula: C4Br4S
InChi Key: AVPWUAFYDNQGNZ-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(Br)S1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)