accessibility menu, dialog, popup

UserName

CAS RN 3934-20-1

IUPAC Name: 2,4-dichloropyrimidine
Synonyms: 2,4-dichloropyrimidine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: BTTNYQZNBZNDOR-UHFFFAOYSA-N
SMILES: ClC1=CC=NC(Cl)=N1
Boiling Point 
  • (2)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (4)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (7)
  • (2)
  • (4)
  • (3)
  • (6)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)