accessibility menu, dialog, popup

UserName

CAS RN 393-77-1

IUPAC Name: 2,6-dinitro-4-(trifluoromethyl)phenol
Synonyms: 2,6-dinitro-4-(trifluoromethyl)phenol
Molecular Weight (g/mol): 252.11
Molecular Formula: C7H3F3N2O5
InChi Key: FXZGYEWQIGIFMC-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results