accessibility menu, dialog, popup

UserName

CAS RN 39267-05-5

H3CNNClCl

CAS RN 39267-05-5

IUPAC Name: 2,3-dichloro-6-methylquinoxaline
Synonyms: 2,3-dichloro-6-methylquinoxaline
Molecular Weight (g/mol): 213.06
Molecular Formula: C9H6Cl2N2
InChi Key: UWSAFTDEEVGSAC-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(Cl)C(Cl)=NC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results