missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-methylquinoxaline, TCI America™

Catalog No. D32705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32705G Supplier TCI America Supplier No. D32705G
Only null left

Chemical Identifiers

CAS 39267-05-5
Molecular Formula C9H6Cl2N2
Molecular Weight (g/mol) 213.06
MDL Number MFCD00270819
InChI Key UWSAFTDEEVGSAC-UHFFFAOYSA-N
PubChem CID 560641
IUPAC Name 2,3-dichloro-6-methylquinoxaline
SMILES CC1=CC=C2N=C(Cl)C(Cl)=NC2=C1

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 213.06
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-6-methylquinoxaline