accessibility menu, dialog, popup

UserName

CAS RN 3895-92-9

IUPAC Name: 17,18-dimethoxy-21-methyl-5,7-dioxa-21-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁴,¹⁹]henicosa-1(13),2,4(8),9,11,14,16,18,20-nonaen-21-ium chloride
Synonyms: chelerythrine chloride
Molecular Weight (g/mol): 383.83
Molecular Formula: C21H18ClNO4
InChi Key: WEEFNMFMNMASJY-UHFFFAOYSA-M
SMILES: [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results