missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chelerythrine chloride, 99+%
Description
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A natural benzophenanthridine alkaloid
- Inhibits protein kinase C
- Induces apoptosis in leukemia cells
Specifications
Specifications
| Color | Yellow to Orange |
| Quantity | 25 mg |
| Merck Index | 14,2051 |
| Solubility Information | Soluble in DMSO at 2mg/ml |
| IUPAC Name | 17,18-dimethoxy-21-methyl-5,7-dioxa-21-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁴,¹⁹]henicosa-1(13),2,4(8),9,11,14,16,18,20-nonaen-21-ium chloride |
| Formula Weight | 383.82 |
| Percent Purity | ≥99% |
| Physical Form | Solid |
| Chemical Name or Material | Chelerythrine chloride |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 223-444-9 |
| RTECSNumber | FL9200000 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only