accessibility menu, dialog, popup

UserName

CAS RN 38896-30-9

H3COON

CAS RN 38896-30-9

IUPAC Name: methyl quinoline-6-carboxylate
Synonyms: methyl quinoline-6-carboxylate
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: XSRWQTDEIOHXSL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2N=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results