missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Quinolinecarboxylate 97.0+%, TCI America™

Catalog No. M16875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16875G 5 g
M168725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16875G Supplier TCI America Supplier No. M16875G
Only null left

Chemical Identifiers

CAS 38896-30-9
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00956766
InChI Key XSRWQTDEIOHXSL-UHFFFAOYSA-N
Synonym methyl 6-quinolinecarboxylate, 6-quinolinecarboxylic acid, methyl ester, methylquinoline-6-carboxylate, 6-quinolinecarboxylic acid methyl ester, methyl 6-quinoline carboxylate, quinoline-6-carboxylic acid methyl ester, pubchem19578, maybridge1_005415, acmc-209j2g, ksc536k0t
PubChem CID 736812
IUPAC Name methyl quinoline-6-carboxylate
SMILES COC(=O)C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point 85°C
Quantity 5 g
Formula Weight 187.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Quinolinecarboxylate