accessibility menu, dialog, popup

UserName

CAS RN 3868-03-9

IUPAC Name: 3,8,9-trioxatricyclo[4.2.1.0²,⁴]nonan-5-ol
Synonyms: 3,8,9-trioxatricyclo[4.2.1.0²,⁴]nonan-5-ol
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: RXDFVNWKKAAOSK-UHFFFAOYNA-N
SMILES: OC1C2OC2C2OCC1O2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results