missing translation for 'onlineSavingsMsg'
Learn More

1,6:2,3-Dianhydro-beta-D-mannopyranose 98.0+%, TCI America™

Catalog No. D4372200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4372200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4372200MG Supplier TCI America Supplier No. D4372200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3868-03-9
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD18643031
InChI Key RXDFVNWKKAAOSK-UHFFFAOYNA-N
Synonym 1,6:2,3-Dianhydro-beta-D-mannose
PubChem CID 11205814
SMILES C1C2C(C3C(O3)C(O1)O2)O

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 200 mg
Formula Weight 144.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6:2,3-Dianhydro-beta-D-mannopyranose