accessibility menu, dialog, popup

UserName

NaNaNaOHNNNHSOOOSOOOSOOO(E/Z)

CAS RN 3861-73-2

IUPAC Name: trisodium 4-hydroxy-5-{2-[4-(phenylamino)-5-sulfonatonaphthalen-1-yl]diazen-1-yl}naphthalene-2,7-disulfonate
Synonyms: trisodium 4-hydroxy-5-{2-[4-(phenylamino)-5-sulfonatonaphthalen-1-yl]diazen-1-yl}naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 695.57
Molecular Formula: C26H16N3Na3O10S3
InChi Key: ADGGJQPKBDIZMT-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OC1=C2C(=CC(=C1)S([O-])(=O)=O)C=C(C=C2N=NC1=C2C=CC=C(C2=C(NC2=CC=CC=C2)C=C1)S([O-])(=O)=O)S([O-])(=O)=O
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results