missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Blue 92, TCI America™
Specifications
Specifications
| Formula Weight | 695.57 |
| Color | Black/Purple |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Acid Blue 92 |
Safety and Handling
| EINECSNumber | (5)-1657 |
| RTECSNumber | QJ6300000 |
| TSCA | Yes |