accessibility menu, dialog, popup

UserName

NaNaOOIOIIOI

CAS RN 386-17-4

IUPAC Name: disodium 2-[(3,5-diiodo-4-oxidophenyl)(3,5-diiodo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
Synonyms: disodium 2-[(3,5-diiodo-4-oxidophenyl)(3,5-diiodo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
Molecular Weight (g/mol): 865.88
Molecular Formula: C20H8I4Na2O4
InChi Key: CWHBCTLVWOCMPQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C(C1=CC(I)=C([O-])C(I)=C1)=C1C=C(I)C(=O)C(I)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results