missing translation for 'onlineSavingsMsg'
Learn More

3',3″,5',5″-Tetraiodophenolphthalein 95.0+%, TCI America™

Catalog No. T01265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01265G 5 g
T012625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01265G Supplier TCI America Supplier No. T01265G

Chemical Identifiers

CAS 386-17-4
Molecular Formula C20H10I4O4
Molecular Weight (g/mol) 821.914
MDL Number MFCD00001614
InChI Key FWQKRBDABCRWKV-UHFFFAOYSA-N
Synonym Iodophene, Iodophthalein, Tetraiodophthalein
PubChem CID 67846
IUPAC Name 3,3-bis(4-hydroxy-3,5-diiodophenyl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC2(C3=CC(=C(C(=C3)I)O)I)C4=CC(=C(C(=C4)I)O)I

Specifications

Color Yellow
Quantity 5 g
Formula Weight 821.91
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,3′′,5′,5′′-Tetraiodophenolphthalein
RTECSNumber SM8397000
TSCA No