accessibility menu, dialog, popup

UserName

CAS RN 384821-19-6

IUPAC Name: methyl[(1-methyl-1H-imidazol-5-yl)methyl]amine
Synonyms: methyl[(3-methylimidazol-4-yl)methyl]amine
Molecular Weight (g/mol): 125.18
Molecular Formula: C6H11N3
InChi Key: YZCXLAHTHBVYGB-UHFFFAOYSA-N
SMILES: CNCC1=CN=CN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results