missing translation for 'onlineSavingsMsg'
Learn More

N-Méthyl-N-[(1-méthyl-1H-imidazol-5-yl)méthyl]amine, 97%, Thermo Scientific™

Numéro de catalogue. CC23646CB
Modifier l'affichage
Click to view available options
Quantité:
250 mg
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC23646CB 250 mg Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC23646CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC23646CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 384821-19-6
Formule moléculaire C6H11N3
Poids moléculaire (g/mol) 125.175
Numéro MDL MFCD07772801
Clé InChI YZCXLAHTHBVYGB-UHFFFAOYSA-N
Synonyme n-methyl-n-1-methyl-1h-imidazol-5-yl methyl amine, methyl-3-methyl-3h-imidazol-4-ylmethyl-amine, 1h-imidazole-5-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-imidazol-5-yl methanamine, methyl 3-methylimidazol-4-yl methyl amine, 1-methyl-5-methylamino methyl-1h-imidazole, methyl 1-methyl-1h-imidazol-5-yl methyl amine, n-methyl-1-3-methylimidazol-4-yl methanamine, n,3-dimethyl-3h-imidazole-4-methanamine, 1h-imidazole-5-methanamine,n,1-dimethyl
PubChem CID 7164555
Nom de l’IUPAC N-méthyl-1-(3-méthylimidazol-4-yl)méthanamine
SOURIRES CNCC1=CN=CN1C

Spécifications

Nom ou substance chimique N-Methyl-N-[(1-methyl-1H-imidazol-5-yl)methyl]amine
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Bouteille en verre ambrée
Quantité 250 mg
Poids de formule 125.17
Pourcentage de pureté 90%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.