accessibility menu, dialog, popup

UserName

CAS RN 383177-54-6

IUPAC Name: 2-(pyridin-4-yl)ethan-1-ol hydrochloride
Synonyms: 4-pyridineethanol hydrochloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: SUDVBWMNUVBMHV-UHFFFAOYSA-N
SMILES: Cl.OCCC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)