missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineethanol Hydrochloride 98.0+%, TCI America™

Catalog No. P12935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12935G 5 g
P129325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12935G Supplier TCI America Supplier No. P12935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 383177-54-6
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD01321183
InChI Key SUDVBWMNUVBMHV-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride
PubChem CID 23090609
IUPAC Name 2-(pyridin-4-yl)ethan-1-ol hydrochloride
SMILES Cl.OCCC1=CC=NC=C1

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 159.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridineethanol Hydrochloride