accessibility menu, dialog, popup

UserName

NH2NHNNHON

CAS RN 38304-91-5

IUPAC Name: 6-amino-2-imino-4-(piperidin-1-yl)-1,2-dihydropyrimidin-1-ol
Synonyms: minoxidil minoxidilum rogaine extra strength (for men) men rogaine rogaine for men minoxidil preparation rogaine for women men's rogaine minoxidil (product) minoxidil (for women)
Molecular Weight (g/mol): 209.25
Molecular Formula: C9H15N5O
InChi Key: ZIMGGGWCDYVHOY-UHFFFAOYSA-N
SMILES: NC1=CC(=NC(=N)N1O)N1CCCCC1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)