accessibility menu, dialog, popup

UserName

CH3SOOOF

CAS RN 383-50-6

IUPAC Name: 2-fluoroethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-fluoroethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 218.24
Molecular Formula: C9H11FO3S
InChi Key: XNRDLSNSMTUXBV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCF
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results