missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. F10565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10565G 5 g
F10561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10565G Supplier TCI America Supplier No. F10565G
Only null left

Chemical Identifiers

CAS 383-50-6
Molecular Formula C9H11FO3S
Molecular Weight (g/mol) 218.24
MDL Number MFCD01658061
InChI Key XNRDLSNSMTUXBV-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid 2-Fluoroethyl Ester, 2-Fluoroethyl Tosylate
PubChem CID 256019
IUPAC Name 2-fluoroethyl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCF

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 218.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoroethyl p-Toluenesulfonate