CAS RN 3827-49-4
IUPAC Name:
2-chloro-5-fluorophenol
Synonyms:
2-chloro-5-fluorophenol
Molecular Weight (g/mol):
146.55
Molecular Formula:
C6H4ClFO
InChi Key:
CMQOZIKIOASEIN-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=CC=C1Cl
1
–
2
of
2
results
1
Promotions Available
2-Chloro-5-fluorophenol, 98%, Thermo Scientific™
2-Chloro-5-fluorophenol, 98%, Thermo Scientific™
CAS: 3827-49-4 Molecular Formula: C6H4ClFO Molecular Weight (g/mol): 146.55 MDL Number: MFCD00042524 InChI Key: CMQOZIKIOASEIN-UHFFFAOYSA-N Synonym: 2-chloro-5-fluoro-phenol,pubchem2146,2-chloro-5-fluorphenol,acmc-209iz2,ksc226k0t,attercop-chm at137231 PubChem CID: 2724522 IUPAC Name: 2-chloro-5-fluorophenol SMILES: OC1=CC(F)=CC=C1Cl
2
Promotions Available
2-Chloro-5-fluorophenol, 98%
2-Chloro-5-fluorophenol, 98%
CAS: 3827-49-4 Molecular Formula: C6H4ClFO Molecular Weight (g/mol): 146.55 MDL Number: MFCD00042524 InChI Key: CMQOZIKIOASEIN-UHFFFAOYSA-N Synonym: 2-chloro-5-fluoro-phenol,pubchem2146,2-chloro-5-fluorphenol,acmc-209iz2,ksc226k0t,attercop-chm at137231 PubChem CID: 2724522 IUPAC Name: 2-chloro-5-fluorophenol SMILES: OC1=CC(F)=CC=C1Cl