missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorophenol, 98%

Catalog No. AAH2856706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2856706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2856706 Supplier Thermo Scientific Chemicals Supplier No. H2856706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3827-49-4 | C6H4ClFO | 146.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3827-49-4
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00042524
InChI Key CMQOZIKIOASEIN-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231
PubChem CID 2724522
IUPAC Name 2-chloro-5-fluorophenol
SMILES OC1=CC(F)=CC=C1Cl

Specifications

Boiling Point 183°C to 185°C
Odor Phenol-like
Refractive Index 1.525
Quantity 5 g
UN Number UN3265
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-fluorophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only