accessibility menu, dialog, popup

UserName

CAS RN 3819-88-3

IUPAC Name: 1-fluoro-3-iodo-5-nitrobenzene
Synonyms: 1-fluoro-3-iodo-5-nitrobenzene
Molecular Weight (g/mol): 267
Molecular Formula: C6H3FINO2
InChi Key: MXPYCSFCKXSPAB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC(I)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)